missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Petri Dish Platinum Electrode Chamber

Catalog No. BTX450504
Change view
Click to view available options
Size (Metric) Gap:
5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap
BTX450504 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX450504 Supplier BTX™ Supplier No. 450504

Petri Dish Platinum Electrode Chamber for ex vivo tissues gene or drug delivery.

  • Simple and easy to use
  • Available in 5 and 15 mm
  • Reusable, lab-grade Pyrex glass

Specifications

For Use With (Application) For ex Vivo Tissues Gene or Drug Delivery Using Large or Uniquely Shapped Tissue Samples
For Use With (Equipment) ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001
Material Glass, Platinum
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Petri Dish Platinum Electrode Chamber, 5 mm gap
Depth (English) 0.12 in.
Depth (Metric) 3 mm
Includes Petri Dish Electrode Chamber Only
Length (English) 0.31 in.
Length (Metric) 8 mm
Size (English) Gap 0.2 in.
Size (Metric) Gap 5 mm
Width (English) 0.2 in.
Width (Metric) 5 mm
Sterility Non-sterile
Connection Type Banana Plug to Micrograbber
Pulse Form Square Wave
Show More Show Less