Learn More
Agilent BioTek Petri Dish Adapter
Petri Dish Adapter for use with Lionheart FX, Synergy Neo2, Cytation 5, Cytation Multi-Mode, Cytation 1, Synergy H1, Synergy HTX, and Synergy LX.
Supplier:  Agilent BioTek 02749
Catalog No. BT02749
Only null left
Explore available promotions
Description
Description
  • For use with 60 mm petri/culture dish
  • Both positions compatible with imaging up to 20x magnification
  • Position 1 compatible with 40x and 60x magnification
Specifications
Specifications
Lionheart FX, Synergy Neo2, Cytation 5, Cytation Multi-Mode, Cytation 1, Synergy H1, Synergy HTX, Synergy LX
Product Suggestions
Product Suggestions