missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pesticides 1 - WP, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-64-1 |
| Flash Point | -17°C - closed cup - Solvent |
| Type | Proficiency Testing Material |
| Quantity | 1.2 mL |