missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Triflumuron, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 64628-44-0 |
| Physical Form | Neat |
| Linear Formula | C15H10 ClF3N2O3 |
| MDL Number | MFCD00072496 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 358.70 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH