missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Triflumizole, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 68694-11-1 |
| Physical Form | Neat |
| Linear Formula | C15H15 ClF3N3 O |
| MDL Number | MFCD13152032 |
| Synonym | (E)-4-Chloro-alpha,alpha,alpha-trifluoro-N-(1-imidazol-1-yl-2-propoxyethylidene)-o-toluidine; 1-{1-{[4-Chloro-2-(trifluoromethyl)phenyl]imino}-2-propoxyethyl}-1 H-imidazole |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 345.75 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH