missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Trifloxystrobin Metabolite CGA 321113, MilliporeSigma™ Supelco™

Catalog No. 111018280
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018280 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018280 Supplier Merck Emd Millipore Supplier No. 3451810MG
Agriculture

Specifications

Product Line PESTANAL
CAS 252913-85-2
Linear Formula C19H17F3N2O4
Synonym (E)-alpha-Methoxyimino-2-[(E)-1-(3-trifluoromethylphenyl)ethylidenaminooxymethyl]phenylacetic acid; Trifloxystrobin acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 394.34
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH