missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Trietazine, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 1912-26-1 |
| Physical Form | Neat |
| Linear Formula | C9H16 ClN5 |
| MDL Number | MFCD00128251 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 229.71 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH