missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Trichlorfon, MilliporeSigma™ Supelco™

Catalog No. 111020034
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020034 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020034 Supplier Merck Emd Millipore Supplier No. 4569850MGR
Agriculture

Specifications

Product Line PESTANAL
CAS 533-50-6
Physical Form Neat
Linear Formula C4H8 Cl3O4 P
MDL Number MFCD00055272
Synonym (2,2,2-Trichloro-1-hydroxyethyl)phosphonic acid dimethyl ester; Metrifonate
Type Analytical Standard
Quantity 50 mg
Formula Weight 257.44
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH