missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Triazophos, MilliporeSigma™ Supelco™

Catalog No. 111019774
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019774 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019774 Supplier Merck Emd Millipore Supplier No. 4569650MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 24017-47-8
Physical Form Neat
Linear Formula C12H16N3O3 PS
MDL Number MFCD00055327
Type Analytical Standard
Quantity 50 mg
Formula Weight 313.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH