missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Triaziflam, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 131475-57-5 |
| Product Line | PESTANAL |
| Physical Form | Neat |
| Linear Formula | C17H24 FN5 O |
| MDL Number | MFCD28009650 |
| Synonym | (RS)-N-[2-(3,5-Dimethylphenoxy)-1-methylethyl]-6-(1-fluoro-1-methylethyl)-1,3,5-triazine-2,4-diamine |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 333.40 |
| Percent Purity | ≥98% (HPLC) |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH