missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Triazamate, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018183
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018183 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018183 Supplier Merck Emd Millipore Supplier No. 68723100MG

Triazamate is a carbamoyl triazole compound, widely used as an insecticide and its mode of action involves the inhibition of cholinesterase in insect pests.

Specifications

CAS 112143-82-5
Product Line PESTANAL
Flash Point 189°C
Physical Form Neat
Linear Formula C13H22N4O3 S
MDL Number MFCD01632354
Melting Point 52°C to 56°C
Synonym Ethyl (3-tert-butyl-1-dimethylcarbamoyl-1 H-1,2,4-triazol-5-ylthio)acetate; Ethyl 2-{{1-[(dimethylamino)carbonyl]-3-(1,1-dimethylethyl)-1 H-1,2,4-triazol-5-yl}thio}acetate
Type Analytical Standard
Quantity 100 mg
Formula Weight 314.40
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) carbon content, 48.66-50.65%
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH