missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Topramezone, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 210631-68-8 |
| Physical Form | Neat |
| Linear Formula | C16H17N3O5 S |
| MDL Number | MFCD09953760 |
| Synonym | [3-(4,5-Dihydro-3-isoxazolyl)-2-methyl-4-(methylsulfonyl)phenyl](5-hydroxy-1-methyl-1 H-pyrazol-4-yl)methanone |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 363.39 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH