missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Thiocyclam hydrogenoxalate, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 31895-22-4 |
| Flash Point | >130°C |
| Physical Form | Neat |
| Linear Formula | C5H11 NS3 · C2H2O4 |
| MDL Number | MFCD00211369 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 271.38 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH