missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Thifluzamide, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018041
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018041 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018041 Supplier Merck Emd Millipore Supplier No. 49792100MG
Only null left

Thifluzamide is a fungicide used in controlling a variety of fungal diseases in crops. It is a succinate dehydrogenase inhibitor and its mode of action involves, interference with succinate ubiquinone reductase in the mitochondrial electron transport chain of fungi.

Agriculture

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line PESTANAL
CAS 130000-40-7
Physical Form Neat
Linear Formula C13H6 Br2F6N2O2 S
MDL Number MFCD03428071
Synonym 2 ′,6 ′-Dibromo-2-methyl-4 ′-trifluoromethoxy-4-trifluoromethyl-1,3-thiazole-5-carboxanilide; N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-5-thiazolecarboxamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 528.06
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH