missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Tetramethrin, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 7696-12-0 |
| Physical Form | Neat |
| Linear Formula | C19H25 NO4 |
| MDL Number | MFCD00084635 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 331.41 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH