missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Tetradifon, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 116-29-0 |
| Physical Form | Neat |
| Linear Formula | C12H6 Cl4O2 S |
| MDL Number | MFCD00018127 |
| Synonym | 2,4,4 ′,5-Tetrachlorodiphenyl sulfone |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 356.05 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH