missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Terbumeton-desethyl, MilliporeSigma™ Supelco™

Catalog No. 111013857
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013857 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013857 Supplier Merck Emd Millipore Supplier No. 36514250MG

Terbumeton-desethyl is a major degradation product of terbumeton, a methoxytriazine herbicide, which is found to be a persistent environmental contaminant.

Agriculture

Specifications

Product Line PESTANAL
CAS 30125-64-5
Flash Point >100°C
Physical Form Neat
Linear Formula C8H15N5 O
MDL Number MFCD00144060
Synonym 2-Amino-4-tert-butylamino-6-methoxy-1,3,5-triazine; Desethylterbumetone
Type Analytical Standard
Quantity 250 mg
Formula Weight 197.24
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH