missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Terbufos, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 13071-79-9 |
| Physical Form | Neat |
| Linear Formula | C9H21O2 PS3 |
| MDL Number | MFCD00055355 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 288.43 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH