missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Tebuthiuron, MilliporeSigma™ Supelco™

Catalog No. 111019484
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019484 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019484 Supplier Merck Emd Millipore Supplier No. 45671250MG
Only null left

Tebuthiuron is a thiadiazolylurea derivative and a root active, soil applied herbicide, effectively used for woody plant control.

Agriculture

Specifications

Product Line PESTANAL
CAS 34014-18-1
Physical Form Neat
Linear Formula C9H16N4 OS
MDL Number MFCD00078732
Type Analytical Standard
Quantity 250 mg
Formula Weight 228.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH