missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Tebuconazole-(trimethyl-13C3), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018025
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018025 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018025 Supplier Merck Emd Millipore Supplier No. 432935MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 1313734-83-6
Linear Formula 13C3C13H22 ClN3 O
MDL Number MFCD19687036
Type Analytical Standard
Quantity 5 mg
Formula Weight 310.80
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH