missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ TBTC, MilliporeSigma™ Supelco™

Catalog No. 111020287
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020287 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020287 Supplier Merck Emd Millipore Supplier No. 45713250MG
Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 1461-22-9
Vapor Pressure <0.01 mmHg (20°C)
Density 1.2 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula [CH3(CH2)3]3 SnCl
MDL Number MFCD00000521
Boiling Point 171°C to 173°C (25 mmHg)
Synonym Tributyltin chloride; TBTC; Tributylchlorotin
Type Analytical Standard
Quantity 250 mg
Formula Weight 325.51
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH