missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Sulprofos, MilliporeSigma™ Supelco™

Catalog No. 111020198
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020198 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020198 Supplier Merck Emd Millipore Supplier No. 45665250MG

Sulprofos is a selective organophosphorus insecticide, primarily used in cotton cultivation.

Agriculture

Specifications

Product Line PESTANAL
CAS 35400-43-2
Physical Form Neat
Linear Formula C12H19O2 PS3
MDL Number MFCD00078721
Synonym Sulprophos
Type Analytical Standard
Quantity 250 mg
Formula Weight 322.45
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH