missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Spirotetramat, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 203313-25-1 |
| Physical Form | Neat |
| Linear Formula | C21H27 NO5 |
| Synonym | cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5 ]dec-3-en-4-yl ethyl carbonate; cis-4-(Ethoxycarbonyloxy)-8-methoxy-3-(2,5-xylyl)-1-azaspiro[4.5 ]dec-3-en-2-one |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 373.44 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH