missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Spirotetramat Metabolite BYI08330 enol-glucoside, MilliporeSigma™ Supelco™

Catalog No. 111018086
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018086 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018086 Supplier Merck Emd Millipore Supplier No. 3248710MG
Only null left

Spirotetramat Metabolite BYI08330 enol-glucoside is one of the major metabolites of spirotetramat in plants.

Agriculture

Specifications

Product Line PESTANAL
CAS 1172614-86-6
Physical Form Neat
Linear Formula C24H33 NO8
Synonym cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro [4.5 ]dec-3-en-4-yl beta-D-glucopyranoside
Type Analytical Standard
Quantity 10 mg
Formula Weight 463.52
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH