missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Spirotetramat Metabolite BYI08330-cis-keto-hydroxy, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 1172134-11-0 |
| Physical Form | Neat |
| Linear Formula | C18H23 NO4 |
| Synonym | cis-3-(2,5-Dimethylphenyl)-3-hydroxy-8-methoxy-1-azaspiro[4.5 ]decane-2,4-dione |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 317.38 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH