missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Spirodiclofen, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 148477-71-8 |
| Physical Form | Neat |
| Linear Formula | C21H24 Cl2O4 |
| MDL Number | MFCD04972571 |
| Synonym | 3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5 ]dec-3-en-4-yl) 2,2-dimethylbutyrate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 411.32 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH