missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Simazine, MilliporeSigma™ Supelco™

Catalog No. 111020170
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020170 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020170 Supplier Merck Emd Millipore Supplier No. 32059250MG

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 122-34-9
Flash Point 100°C - Closed Cup
Physical Form Neat
Linear Formula C7H12 ClN5
MDL Number MFCD00023174
Synonym 2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine
Type Analytical Standard
Quantity 250 mg
Formula Weight 201.66
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH