missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Simazine-d10, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 220621-39-6 |
| Physical Form | Neat |
| Linear Formula | C7H2D10 ClN5 |
| MDL Number | MFCD01632307 |
| Synonym | 2,4-Bis(pentadeuteroethylamino)-6-chloro-1,3,5-triazine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 211.72 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH