missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ S-Metolachlor Metabolite CGA 50720, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 152019-74-4 |
| Physical Form | Neat |
| Linear Formula | C11H13 NO3 |
| MDL Number | MFCD10018455 |
| Synonym | N-(2-Ethyl-6-methylphenyl)-oxalamic acid; [(2-Ethyl-6-methylphenyl)amino]oxo-acetic acid |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 207.23 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH