missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Quizalofop-P-tefuryl Solution, 100 ng/L in Acetonitrile, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 200509-41-7 |
| Linear Formula | C22H21 ClN2O5 |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 428.87 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 ng/μL in acetonitrile |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH