missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Quizalofop-P-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111019469
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019469 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019469 Supplier Merck Emd Millipore Supplier No. 34074100MG
Only null left

Quizalofop-p-ethyl is a post-emergence herbicide, which belongs to the aryloxyphenoxypropionate group of herbicides. It commonly finds applications in annual and perennial weed control management.

Agriculture

Specifications

Product Line PESTANAL
CAS 100646-51-3
Flash Point >100°C - Closed Cup
Physical Form Neat
Linear Formula C19H17 ClN2O4
MDL Number MFCD09028086
Synonym DPX-Y6202; Ethyl (R)-2-[4-(6-chloro-2-quinoxalyloxy)phenoxy]propionate; Quinofop-ethyl
Type Analytical Standard
Quantity 100 mg
Formula Weight 372.80
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH