missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Quinoclamine, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Chemical Name or Material | Quinoclamine |
| CAS | 2797-51-5 |
| Product Line | PESTANAL |
| Physical Form | Neat |
| Linear Formula | C10H6 ClNO2 |
| Technique | HPLC |
| MDL Number | MFCD00001680 |
| Synonym | 2-Amino-3-chloro-1,4-naphthoquinone |
| Type | Analytical Standard |
| Molecular Weight (g/mol) | 207.61 |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH