missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Pyrimethanil, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 53112-28-0 |
| Physical Form | Neat |
| Linear Formula | C12H13N3 |
| MDL Number | MFCD00172113 |
| Synonym | 2-Anilino-4,6-dimethylpyrimidine |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 199.25 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH