missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Pyridafol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014142
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014142 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014142 Supplier Merck Emd Millipore Supplier No. 9297425MG

Specifications

CAS 40020-01-7
Product Line PESTANAL
Physical Form Neat
Linear Formula C10H7 ClN2 O
MDL Number MFCD00143498
Synonym 3-Chloro-5-hydroxy-6-phenylpyridazine; 6-Chloro-3-phenyl-4-pyridazinol
Type Analytical Standard
Quantity 25 mg
Formula Weight 206.63
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH