missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Pyraflufen-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111018055
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018055 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018055 Supplier Merck Emd Millipore Supplier No. 3534610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pyraflufen-ethyl is a herbicide which belongs to the phenylpyrazole class. It is used in controlling annual and perennial broadleaf weeds. The mode of action involves inhibiting the enzyme activity of protoporphyrinogen oxidase (Protox).

Agriculture

Specifications

Product Line PESTANAL
CAS 129630-19-9
Physical Form Neat
Linear Formula C15H13 Cl2F3N2O4
MDL Number MFCD07363861
Synonym Ethyl [2-chloro-5-(4-chloro-5-difluoromethoxy-1-methylpyrazol-3-yl)-4-fluorophenoxy]acetate
Type Analytical Standard
Quantity 10 mg
Formula Weight 413.18
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH