missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Pymetrozin, MilliporeSigma™ Supelco™

Catalog No. 111012821
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012821 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012821 Supplier Merck Emd Millipore Supplier No. 46119250MGR
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 123312-89-0
Flash Point >230°C
Physical Form Neat
Linear Formula C10H11N5 O
MDL Number MFCD01632346
Type Analytical Standard
Quantity 250 mg
Formula Weight 217.23
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH