missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Prothioconazole, MilliporeSigma™ Supelco™

Catalog No. 111018418
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018418 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018418 Supplier Merck Emd Millipore Supplier No. 34232100MG

Prothioconazole is a triazolinethione derivative, which can be used as a fungicide in order to inhibit the activity of demethylase enzyme.(1) It can be used in the treatment of infection in crops like wheat, caused by Mycosphaerella graminicola, a plant-pathogenic fungus.

Agriculture

Specifications

Product Line PESTANAL
CAS 178928-70-6
Physical Form Neat
Linear Formula C14H15 Cl2N3 OS
Synonym 2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3 H-1,2,4-triazole-3-thione
Type Analytical Standard
Quantity 100 mg
Formula Weight 344.26
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH