missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Prothioconazole-desthio, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 120983-64-4 |
| Physical Form | Neat |
| Linear Formula | C14H15 Cl2N3 O |
| Synonym | alpha-(1-Chlorocyclopropyl)-alpha-[(2-chlorophenyl)methyl]-1 H-1,2,4-triazole-1-ethanol |
| Type | Analytical Standard |
| Quantity | 20 mg |
| Formula Weight | 312.19 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH