missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Propylene thiourea, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 2122-19-2 |
| Physical Form | Neat |
| Linear Formula | C4H8N2 S |
| MDL Number | MFCD00121569 |
| Synonym | 4-Methyl-2-imidazolidinethione |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 116.18 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH