missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Propineb, MilliporeSigma™ Supelco™

Catalog No. 111012754
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012754 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012754 Supplier Merck Emd Millipore Supplier No. 45643250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Propineb belongs to the dithiocarbamates (DTCs) class of fungicides, widely used in agriculture and horticulture. It has an additional advantage of low production cost, broad-spectrum of chemical and biological reactivity.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 12071-83-9
Physical Form Neat
Linear Formula [C5H8N2S4 Zn]n
MDL Number MFCD16661267
Synonym Zinc propylenebis(dithiocarbamate)
Type Analytical Standard
Quantity 250 mg
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH