missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Propazine, MilliporeSigma™ Supelco™

Catalog No. 111020194
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020194 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020194 Supplier Merck Emd Millipore Supplier No. 45640250MG

Propazine is a triazine herbicide, used in controlling broad-leaved weeds and annual grasses in sweet sorghum.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 139-40-2
Physical Form Neat
Linear Formula C9H16 ClN5
MDL Number MFCD00047341
Synonym 2,4-Bis(isopropylamino)-6-chloro-1,3,5-triazine
Type Analytical Standard
Quantity 250 mg
Formula Weight 229.71
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH