missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Propamocarb, MilliporeSigma™ Supelco™

Catalog No. 111013681
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013681 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013681 Supplier Merck Emd Millipore Supplier No. 45638250MG
Only null left

Propamocarb is a systemic fungicide, widely used for greenhouse vegetables and agricultural commodities, which shows protective action against phycomycetous diseases (Phythium, Phytophthora spp.).

Agriculture

Specifications

Product Line PESTANAL
CAS 24579-73-5
Flash Point >100°C
Physical Form Neat
Linear Formula C9H20N2O2
MDL Number MFCD00078717
Type Analytical Standard
Quantity 250 mg
Formula Weight 188.27
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH