missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Profenofos, MilliporeSigma™ Supelco™

Catalog No. 111020545
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020545 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020545 Supplier Merck Emd Millipore Supplier No. 45632250MG

Profenofos is a broad-spectrum organophosphorus pesticide, commonly used for the control of various insect pests on vegetable crops.

Agriculture

Specifications

Product Line PESTANAL
CAS 41198-08-7
Flash Point 124°C
Physical Form Neat
Linear Formula C11H15 BrClO3 PS
MDL Number MFCD00078736
Type Analytical Standard
Quantity 250 mg
Formula Weight 373.63
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH