missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Procymidone, MilliporeSigma™ Supelco™

Numéro de catalogue. 111013903
Modifier l'affichage
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111013903 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111013903 Fournisseur Merck Emd Millipore Code fournisseur 36640250MGR
Agriculture

Spécifications

Product Line PESTANAL
CAS 3129-91-7
Physical Form Sympa
Linear Formula C13H11 Cl2 NO2
MDL Number MFCD00078740
Type Analytical Standard
Quantity 250 mg
Formula Weight 284.14
Grade Norme analytique

PESTANAL est une marque déposée d’International Sigma-Aldrich GmbH