missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Prochloraz Metabolite BTS44596, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 139542-32-8 |
| Linear Formula | C13H15 Cl3N2O3 |
| MDL Number | MFCD28122615 |
| Synonym | N′-Formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]urea |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 353.63 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH