missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Prochloraz Metabolite BTS40348, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 67747-01-7 |
| Linear Formula | C11H14 Cl3 NO |
| MDL Number | MFCD09751307 |
| Synonym | N-[2-(2,4,6-Trichlorophenoxy)ethyl]propylamine; N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]amine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 282.59 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH