missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Primisulfuron-methyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 86209-51-0 |
| Physical Form | Neat |
| Linear Formula | C15H12F4N4O7 S |
| MDL Number | MFCD00337169 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 468.34 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH