missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Piroctone olamine, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015181
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015181 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015181 Supplier Merck Emd Millipore Supplier No. 51872100MG
Only null left

Piroctone olamine is a pyridone derivative, which is known to have bactericidal effects on gram-positive and gram-negative bacteria and fungicidal effects and hence is a component of many cosmetic products such as anti-dandruff shampoo.

Specifications

CAS 68890-66-4
Product Line PESTANAL
Physical Form Neat
Linear Formula C14H23 NO2 · C2H7 NO
MDL Number MFCD01690792
Synonym 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1 H)-pyridone ethanolammonium salt
Type Analytical Standard
Quantity 100 mg
Formula Weight 298.42
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH