missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Pirimiphos-methyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 29232-93-7 |
| Physical Form | Neat |
| Linear Formula | C11H20N3O3 PS |
| MDL Number | MFCD00055366 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 305.33 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH