missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Pirimicarb, MilliporeSigma™ Supelco™

Catalog No. 111020539
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020539 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020539 Supplier Merck Emd Millipore Supplier No. 45627250MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 23103-98-2
Flash Point >100°C
Physical Form Neat
Linear Formula C11H18N4O2
MDL Number MFCD00055520
Synonym 2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethylcarbamate
Type Analytical Standard
Quantity 250 mg
Formula Weight 238.29
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH